Learn More
Meso Scale Discovery Kidney Injury PNL 1 rat Kit
SDP
Catalog No. 5000082258
Encompass
The Meso Scale Discovery Kidney Injury Panel 1 Rat Kit is a multiplex immunoassay that measures biomarkers of kidney injury and function in rat serum plasma and urine The kit includes a 96-well plate coated with capture antibodies for KIM-1 Clusterin Cystatin C NGAL and Creatinine The assay uses electrochemiluminescence detection and takes under 4 hours to run The kit includes all necessary reagents and calibrators and can process up to 80 samples in duplicate The assay has high sensitivity and a broad dynamic range to quantitate kidney biomarkers This product enables researchers to evaluate kidney toxicity and injury in preclinical rat studies
Catalog No. 50-000-82258 Supplier Meso Scale Discovery Supplier No. K15162C1
The Meso Scale Discovery Kidney Injury Panel 1 Rat Kit is a multiplex immunoassay that measures biomarkers of kidney injury and function in rat serum plasma and urine The kit includes a 96-well plate coated with capture antibodies for KIM-1 Clusterin Cystatin C NGAL and Creatinine The assay uses electrochemiluminescence detection and takes under 4 hours to run The kit includes all necessary reagents and calibrators and can process up to 80 samples in duplicate The assay has high sensitivity and a broad dynamic range to quantitate kidney biomarkers This product enables researchers to evaluate kidney toxicity and injury in preclinical rat studies