Learn More
Meso Scale Discovery Kidney Injury PNL 3 human Kit
SDP
Catalog No. 5000080261
Encompass
The Meso Scale Discovery Kidney Injury Panel 3 (Human) Kit is a multiplex immunoassay that simultaneously measures KIM-1 Clusterin and Cystatin-C in human serum plasma urine and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This product enables sensitive reproducible quantification of kidney injury biomarkers
Catalog No. 50-000-80261 Supplier Meso Scale Discovery Supplier No. K15189D1
The Meso Scale Discovery Kidney Injury Panel 3 (Human) Kit is a multiplex immunoassay that simultaneously measures KIM-1 Clusterin and Cystatin-C in human serum plasma urine and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This product enables sensitive reproducible quantification of kidney injury biomarkers