Learn More
Meso Scale Discovery Kidney Injury PNL 3 human Kit
SDP
Catalog No. 5000082450
Encompass
The Meso Scale Discovery Kidney Injury Panel 3 Human Kit is a multiplex assay for measuring kidney injury biomarkers like KIM-1 Clusterin and Cystatin C in human samples It uses electrochemiluminescent detection for sensitive and accurate quantification The kit includes a 96-well plate with capture and detection antibodies calibrators controls buffers and seals Compatible with MSD instruments it aids in assessing diagnosing and monitoring acute kidney injury
Catalog No. 50-000-82450 Supplier Meso Scale Discovery Supplier No. K15189D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Kidney Injury Panel 3 Human Kit is a multiplex assay for measuring kidney injury biomarkers like KIM-1 Clusterin and Cystatin C in human samples It uses electrochemiluminescent detection for sensitive and accurate quantification The kit includes a 96-well plate with capture and detection antibodies calibrators controls buffers and seals Compatible with MSD instruments it aids in assessing diagnosing and monitoring acute kidney injury