Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProcartaPlex™ Human Kidney Toxicity Panel 1, 11plex

Catalog No. 502557011
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-7011 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-7011 Supplier Invitrogen™ Supplier No. EPX11015855901
Add to Cart
Add to Cart

The ProcartaPlex Human Kidney Toxicity Panel 1 11plex enables nephrotoxicity research by analyzing 11 protein targets in a single well using Luminex xMAP technology.

The ProcartaPlex Human Kidney Toxicity Panel 1 11plex enables nephrotoxicity research by analyzing 11 protein targets in a single well using Luminex xMAP technology. Simultaneous detection of these targets can help to gain more information about kidney functions and nephrotoxicity during drug development studies.

Target list [bead region]: calbindin [37], clusterin (APO-J) [72], GSTA1 (glutathione S-transferase A1) [42], IL-18 [66], IP-10 (CXCL10) [22], KIM-1 (TIM-1/HAVCR1) [53], MCP-1 (CCL2) [51], osteoactivin (GPNMB) [29], RBP4 (retinol-binding protein 4) [33], renin [46]; and VEGF-A [78]

The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels. It complements the ProcartaPlex Human Kidney Toxicity Panel 2 9plex (Cat. No. EPX090-15856-901) that provides 9 additional protein targets in the same research area.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit our ProcartaPlex Immunoassays page for more information and available products.

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 65 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Combinability Non-Combinable
Bead Type Magnetic
Bead Region Calbindin [37], Clusterin (APO-J) [72], GSTA1 (Glutathione S-transferase A1) [42], IL-18 [66], IP-10 (CXCL10) [22], KIM-1 (TIM-1/HAVCR1) [53], MCP-1 (CCL2) [51], Osteoactivin (GPNMB) [29], RBP4 (Retinol-binding protein 4) [33], Renin [46], VEGF-A [78]
Assay Sensitivity See product documentation
Assay Range See product documentation
Format Multiplex Kit
For Use With (Equipment) Luminex™ Instruments
High-throughput Compatibility No
Interassay CV See product documentation
Intraassay CV See product documentation
Product Line ProcartaPlex
Sample Type Urine, Serum, Plasma, Cell Culture Supernatants
Sample Volume Urine, Serum, Plasma: 25 μL, CCS: 50 μL
Content And Storage 1 vial Capture Beads (50x)
1 vial biotinylated Detection Antibody (50x)
2 vials Standard Mix 15 (lyophilized)
2 vials Standard Mix 16 (lyophilized)

Store at 2-8°C.
Shipping Condition Wet Ice
Research Area Toxicology, Immunology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.