Learn More
Vernier KidWind Wind Turbine Hubs
SDPSCI_ED
Catalog No. S16180ND Shop All Vernier Software Products
Change view
Click to view available options
Quantity:
10/Pk.
3/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S16180ND 3/Pk.
S04833ND 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S16180ND Supplier Vernier Supplier No. KWWTH3
Add to Cart
Add to Cart
To turn any DC generator into a wind turbine.
  • Recycled plastic made 12-hole crimping
  • Press fits onto shaft of small DC generators to generate wind power
  • Allows blade angles and/or interchange blade sets to be changed
  • Diameter: 5.7cm (2.25 in.)
  • Max number of dowels: 12
  • Size of shaft hub press fits onto: 2mm
  • Recommended for college and high school
Warranty and Services
5-year limited warranty
Specifications
Product Type Wind Turbine Hubs
Material Recycled Plastic
Quantity 3/Pk.