Learn More
Aviva Systems Biology Corp KIF13B Antibody - N-terminal region (ARP33926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF13B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIF13B. Alternate names: GAKIN, KIAA0639ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55200