Learn More
Aviva Systems Biology Corp KIF15 Antibody - middle region (ARP33934_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33934_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIF15. Alternate names: FLJ25667, HKLP2, KNSL7, NY-BR-62ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-55201