Learn More
Apexbio Technology LLC Kif15-IN-1(Synonyms: Kif15 inhibitor 1, Kif15-IN1, Kif15 inhibitor I, KIF15-IN-1, Kif15 motor protein inhibitor), 2mg, CAS: 672926-32-8.
SDP
Catalog No. 502240064
Encompass_Preferred
Kif15-IN-1 (CAS 672926-32-8) is a small molecule inhibitor targeting kinesin family member 15 (Kif15) also termed kinesin-12 or HKLP2 Kif15 is a kinesin-related motor protein expressed during mitosis crucially involved in the formation and stabilization of bipolar microtubule spindles acting redundantly with KIF11 (Eg5) Inhibition of Kif15 by Kif15-IN-1 results in suppression of tumor cell proliferation highlighting its utility in cell biology research focused on mitotic spindle mechanics and its potential in oncology as a candidate for antiproliferative strategies
This item is not returnable. View return policy
Catalog No. 50-224-0064 Supplier Apexbio Technology LLC Supplier No. B32802
Kif15-IN-1 (CAS 672926-32-8) is a small molecule inhibitor targeting kinesin family member 15 (Kif15) also termed kinesin-12 or HKLP2 Kif15 is a kinesin-related motor protein expressed during mitosis crucially involved in the formation and stabilization of bipolar microtubule spindles acting redundantly with KIF11 (Eg5) Inhibition of Kif15 by Kif15-IN-1 results in suppression of tumor cell proliferation highlighting its utility in cell biology research focused on mitotic spindle mechanics and its potential in oncology as a candidate for antiproliferative strategies