Learn More
Medchemexpress LLC HY-15949 5mg Medchemexpress, Kif15-IN-2 CAS:672926-33-9 Purity:>98%
SDP
Catalog No. 502081845
Encompass_Preferred
Medchemexpress, HY-15949 5mg Kif15-IN-2 CAS:672926-33-9 Kif15-IN-2 is an inhibitor of the mitotic kinesin Kif15, and is used for the research of cellular proliferative diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1845 Supplier Medchemexpress LLC Supplier No. HY159495MG
Medchemexpress, HY-15949 5mg Kif15-IN-2 CAS:672926-33-9 Kif15-IN-2 is an inhibitor of the mitotic kinesin Kif15, and is used for the research of cellular proliferative diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.