Learn More
Aviva Systems Biology Corp KIF1C Antibody - C-terminal region (ARP33913_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33913_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF1C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KIF1C. Alternate names: LTXS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55195