Learn More
ABclonal Technology KIF1C Rabbit pAb
SDP
Catalog No. 5000105544
Encompass_Preferred
The protein encoded by this gene is a member of the kinesin-like protein family. The family members are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. Mutations in this gene are a cause of spastic ataxia 2, autosomal recessive.
Catalog No. 50-001-05544 Supplier ABclonal Technology Supplier No. A15786500UL
The protein encoded by this gene is a member of the kinesin-like protein family. The family members are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. Mutations in this gene are a cause of spastic ataxia 2, autosomal recessive.