Learn More
ABclonal Technology KIF20B Rabbit pAb
SDP
Catalog No. 5000108072
Encompass_Preferred
Enables several functions, including WW domain binding activity, plus-end-directed microtubule motor activity, and protein homodimerization activity. Involved in positive regulation of cell population proliferation and positive regulation of cytokinesis. Located in several cellular components, including microtubule cytoskeleton, midbody, and nuclear lumen.
Catalog No. 50-001-08072 Supplier ABclonal Technology Supplier No. A15360500UL
Enables several functions, including WW domain binding activity, plus-end-directed microtubule motor activity, and protein homodimerization activity. Involved in positive regulation of cell population proliferation and positive regulation of cytokinesis. Located in several cellular components, including microtubule cytoskeleton, midbody, and nuclear lumen.