Learn More
Aviva Systems Biology Corp KIF21A Antibody - N-terminal region (ARP33932_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33932_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF21A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIF21A. Alternate names: FEOM1, CFEOM1, FEOM3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-52937