Learn More
Aviva Systems Biology Corp KIF22 Antibody - C-terminal region (ARP33920_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33920_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KIF22. Alternate names: KID, KNSL4, OBP, OBP-1, OBP-2, SEMDJL2, A-328A3.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-55199