Learn More
Aviva Systems Biology Corp KIF23 Antibody - N-terminal region (ARP33962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIF23. Alternate names: CHO1, KNSL5, MKLP1, MKLP-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55207