Learn More
Aviva Systems Biology Corp KIF25 Antibody - C-terminal region (ARP33925_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33925_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF25. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human KIF25. Alternate names: KNSL3ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-52936