Learn More
Aviva Systems Biology Corp KIF25 Antibody - N-terminal region (ARP33972_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33972_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIF25. Alternate names: KNSL3, MGC163361ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55211