Learn More
Aviva Systems Biology Corp KIF3B Antibody - C-terminal region (ARP33908_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33908_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KIF3B. Alternate names: PARP, PPOL, ADPRT, ADPRT1, PARP-1, ADPRT 1, pADPRT-1, KIF3BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52935