Learn More
Aviva Systems Biology Corp KIF5A Antibody - middle region (ARP33868_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF5A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human KIF5A. Alternate names: NKHC, MY050, SPG10, D12S1889ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55181