Learn More
Aviva Systems Biology Corp KIF5B Antibody - N-terminal region (ARP33903_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33903_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF5B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KIF5B. Alternate names: KNS, KINH, KNS1, UKHCApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-52933