Learn More
Sino Biological Recombinant Human PTC2 (PRKAR1A & RET) Protein (GST Tag), Active
Catalog No. 502573391
Encompass_Preferred
Recombinant human RET/PTC2, the fusion protein [PRKAR1A (1-236)-RET (713-end)], was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3391 Supplier Sino Biological Supplier No. R0219CG50
Recombinant human RET/PTC2, the fusion protein [PRKAR1A (1-236)-RET (713-end)], was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.