Learn More
Aviva Systems Biology Corp KIF5C Antibody - N-terminal region (ARP54736_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54736_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF5C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIF5C. Alternate names: FLJ44735, KIAA0531, KINN, MGC111478, NKHC, NKHC-2, NKHC2ApplicationIP, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61294