Learn More
Aviva Systems Biology Corp KIF9 Antibody - N-terminal region (ARP33966_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33966_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIF9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human KIF9. Alternate names: MGC104186ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55209