Learn More
Aviva Systems Biology Corp KIFAP3 Antibody - middle region (ARP55127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIFAP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIFAP3. Alternate names: FLJ22818, KAP3, SMAP, Smg-GDS, dJ190I16.1, KAP-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53912