Learn More
Aviva Systems Biology Corp KIFC2 Antibody - N-terminal region (ARP33950_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33950_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIFC2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KIFC2. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse
Catalog No. 50-014-55205