Learn More
Aviva Systems Biology Corp KIFC2 Antibody - N-terminal region (ARP33951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIFC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIFC2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast
Catalog No. 50-014-55206