Learn More
Enzo Life Sciences Kifunensine (1 mg). CAS: 109944-15-2
SDP
Catalog No. 502011739
Encompass_Preferred
Inhibits α-mannosidase. Inhibits asparagine-linked oligosaccharide processing. Immunomodulator. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1739 Supplier Enzo Life Sciences Supplier No. BMLS1140001
Inhibits α-mannosidase. Inhibits asparagine-linked oligosaccharide processing. Immunomodulator. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.