Learn More
Cayman Chemical Kifunensine, 109944-15-2, 5 mg
SDP
Supplier:  Cayman Chemical 100094375
Encompass_Preferred
Molecular Formula C8H12N2O6, Purity ≥98%, Formula Weight 232.2, Synonyms: FR900494; A potent and selective inhibitor of class I -mannosidases and serves as a key inhibitor of glycoprotein biosynthesis; inhibits both human ER -1,2-mannosidase I and Golgi class I mannosidases (Golgi -mannosidase IA, IB, and IC) exhibiting Ki values of 130 and 23 nM, respectively; inhibits mung bean -1,2-mannosidase I with an IC50 value of 20-50 nM.
Catalog No. NC9744221
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.