Learn More
Medchemexpress LLC HY-19332 5mg Medchemexpress, Kifunensine CAS:109944-15-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY193325MG
Encompass_Preferred
Medchemexpress, HY-19332 5mg Kifunensine CAS:109944-15-2 Kifunensine, a potent and selective inhibitor of class I α-mannosidases isolated from Actinomycete, prevents α-mannosidases I from trimming mannose residues on glycoproteins. Kifunensine inhibits ERAD[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0848