Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kifunensine, >98%, MP Biomedicals™

Catalog No. icn15995201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15995201 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15995201 Supplier MP Biomedicals, Inc Supplier No. 0215995201
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109944-15-2
Molecular Formula C8H12N2O6
Molecular Weight (g/mol) 232.192
InChI Key OIURYJWYVIAOCW-PQMKYFCFSA-N
Synonym kifunensine, +-kifunensine, kifunensine, kitasatosporia kifunense, 5r,6r,7s,8r,8as-6,7,8-trihydroxy-5-hydroxymethyl hexahydroimidazo 1,2-a pyridine-2,3-dione, kif, 5r,6r,7s,8r,8as-6,7,8-trihydroxy-5-hydroxymethyl-1,5,6,7,8,8a-hexahydroimidazo 1,2-a pyridine-2,3-dione, 5r,6r,7s,8r,8as-hexahydro-6,7,8-trihydroxy-5-hydroxymethyl-imidazo 1,2-a pyridine-2,3-dione, imidazo 1,2-a pyridine-2,3-dione, hexahydro-6,7,8-trihydroxy-5-hydroxymethyl-, 5r-5alpha,6beta,7alpha,8alpha,8aalpha, 2wvz, d0ad9y
PubChem CID 130611
IUPAC Name (5R,6R,7S,8R,8aS)-6,7,8-trihydroxy-5-(hydroxymethyl)-1,5,6,7,8,8a-hexahydroimidazo[1,2-a]pyridine-2,3-dione
SMILES C(C1C(C(C(C2N1C(=O)C(=O)N2)O)O)O)O

Specifications

Color White
Assay Percent Range >98%
Quantity 1 mg
Formula Weight 232.2
Physical Form Powder
Chemical Name or Material Kifunensine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.