Learn More
Abcam Kifunensine, class I alpha-mannosidase inhibitor, 5MG
Catalog No. 502588157
Encompass_Preferred
MW 232.19 Da. Cell-permeable, potent, selective inhibitor of class I α-mannosidases. (IC₅₀ = 20-50 nM for mung bean α-1,2-mannosidase I). Alkaloid originally isolated from the actinomycete Kitasatosporia kifunense. Used to suppress Endoplasmic Reticulum-Associated Degradation (ERAD) via the inhibition of endoplasmic reticulum-associated mannosidase activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-8157 Supplier Abcam Supplier No. AB2151165MG
MW 232.19 Da. Cell-permeable, potent, selective inhibitor of class I α-mannosidases. (IC₅₀ = 20-50 nM for mung bean α-1,2-mannosidase I). Alkaloid originally isolated from the actinomycete Kitasatosporia kifunense. Used to suppress Endoplasmic Reticulum-Associated Degradation (ERAD) via the inhibition of endoplasmic reticulum-associated mannosidase activity.
The product is subject to the following: Abcam Restricted Use Statement