Learn More
Cayman Chemical KIgamIcIn C 2.5mg
SDP
Catalog No. 502982114
Encompass_Preferred
An antitumor antibiotic; active against various strains of S. aureus, including MRSA, as well as M. luteus and B. subtilis (MICs = 0.05-0.2 UG/ml for all); is cytotoxic to PANC-1 cells in nutrient-deprived media
Catalog No. 50-298-2114 Supplier Cayman Chemical Supplier No. 194452.5 MG
An antitumor antibiotic; active against various strains of S. aureus, including MRSA, as well as M. luteus and B. subtilis (MICs = 0.05-0.2 UG/ml for all); is cytotoxic to PANC-1 cells in nutrient-deprived media