Learn More
Cayman Chemical Kijanimicin, Purity: ≥95%, CAS: 78798-08-0, 2.5 mg
SDP
Catalog No. 502982055
Encompass_Preferred
An antibiotic active against a broad spectrum of microorganisms in vitro including P. acnes (MIC = 0.86 µg/ml), B. subtilis (MIC Enterobacter sp. (MIC = 64 µg/ml), Trichophyton sp. (MIC = 17.5 µg/ml), and Microsporum sp. (MIC = 17.5 µg/ml); effective in mice against P. berghei, a protozoan parasite that causes malaria in certain rodents
Catalog No. 50-298-2055 Supplier Cayman Chemical Supplier No. 205862.5 MG
An antibiotic active against a broad spectrum of microorganisms in vitro including P. acnes (MIC = 0.86 µg/ml), B. subtilis (MIC Enterobacter sp. (MIC = 64 µg/ml), Trichophyton sp. (MIC = 17.5 µg/ml), and Microsporum sp. (MIC = 17.5 µg/ml); effective in mice against P. berghei, a protozoan parasite that causes malaria in certain rodents