Learn More
Enzo Life Sciences KillerTRAIL™-Protein (soluble) (human), (recombinant) (BULK) (500µg)
SDP
Supplier:  Enzo Life Sciences ALX201123C500
Encompass_Preferred
Alternative name: Apo-2L, TNFSF 10, CD253. Endotoxin: <10EU/µg protein (LAL test; Bio Whittaker). Purity: ≥98% (SDS-PAGE). Formulation: Liquid. In 20mM HEPES, pH 7.4, containing 150mM sodium chloride, 0.005% Tween 20, 1% sucrose, 1mM DTT. Source: Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide. Specificity: Binds to human and mouse TRAIL receptors and osteoprotegerin (OPG). Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-7925