Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci KillerTRAIL, Soluble Recombinant Protein
SDP

Catalog No. 16835020
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-835-020 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-835-020 Supplier ProSci Supplier No. 903350050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This recombinant proteins is for research use only.

TNF-related apoptosis-inducing ligand (TRAIL; Apo2L;CD253; TNFSF10) is a type II transmembrane protein of about 34kDa. Like most members of the tumor necrosis factor (TNF) superfamily of cytokines TRAIL can be cleaved at the cell surface by metalloproteases to form a soluble molecule. Active TRAIL forms trimers and specifically binds to five distinct receptors: TRAIL-R1 (DR4; Apo2;CD261; TNFRSF10A), TRAIL-R2 (DR5; KILLER; TRICK2A;TRICK2B; CD262; TNFRSF10B), TRAIL-R3 (DcR1;LIT; TRID; CD263; TNFRSF10C), TRAIL-R4 (DcR2;TRUNDD; CD264; TNFRSF10D), and osteoprotegerin (OPG; OCIF; TNFRSF11B). Trimerized TRAIL triggers apoptosis upon ligation of cell surface TRAIL-R1 and/or TRAIL-R2 by inducing the formation of the so-called multiprotein death-inducing signaling complex (DISC).

Specifications

Accession Number P50591
Concentration 0.5 mg/mL
For Use With (Application) Not Available
Gene ID (Entrez) 8743
Quantity 50 μg
Source E. coli
Common Name tumor necrosis factor (ligand) superfamily, member 10
Gene Symbol TNFSF10
Biological Activity Potent biologically active trimer. Induces apoptosis in a concentration range of 10-100 ng/mL Note: Does not require a cross-linking enhancer for its potent biological activity. ED50: 20 ng/mL (BJAB cells). For cell lines that require extensive cross-linking of the TRAIL-Rs for killing (e.g. Jurkat) use SuperKillerTRAILTM (Prod. No. AG-40 T-0002).
Conjugate Unconjugated
Species Human, Mouse
Recombinant Recombinant
Protein Tag His Tag
Sequence The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide.
Content And Storage Stable for at least 1 year after receipt when stored (at -80°C.)
Form Lyophilized
Purity or Quality Grade >98% (SDS-PAGE). Endotoxin level is less than 0.01 EU/ μg purified protein (LAL test; Lonza).
Synonym TRAIL, Apo-2 L, TNFSF10, CD253
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.