Learn More
Eurofins Discoverx KILR CD16 Effector Cells, 1 vial. (97-0007-01)
Supplier:  Eurofins Discoverx 97000701
Encompass
These KILR CD16 Effector Cells are purified human cytotoxic T lymphocytes stably expressing CD16, intended for use as effector cells in antibody-dependent cell-mediated cytotoxicity (ADCC) assays assessing cell death. They can be used in any cytotoxicity assay measuring cell death, including KILR assays as well as other assays such as europium-release assay. The cells are produced in a standardized batch to allow for high levels of assay reproducibility. These purified effector cells are provided as one vial of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kilr.
Catalog No. NC1406939