Learn More
Eurofins Discoverx KILR Detection Kit, 2,500 dp (96-well). (97-0001L)
Catalog No. 502004546
Encompass
The KILR Detection Kit is a detection kit intended for use with KILR cell lines or pools when running ADCC or other lysis-based assays. This kit contains exogenous EA as well as EFC substrate to allow for detection of lysis-based release of ProLabel-tagged KILR reporter protein. This kit contains all components required to assay 25 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-97-0001l.
Catalog No. 50-200-4546 Supplier Eurofins Discoverx Supplier No. 970001L
The KILR Detection Kit is a detection kit intended for use with KILR cell lines or pools when running ADCC or other lysis-based assays. This kit contains exogenous EA as well as EFC substrate to allow for detection of lysis-based release of ProLabel-tagged KILR reporter protein. This kit contains all components required to assay 25 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-97-0001l.