Learn More
Eurofins Discoverx KILR Detection Kit, 200 dp (96-well). (97-0001S)
Catalog No. NC1000669
Encompass
The KILR Detection Kit is a detection kit intended for use with KILR cell lines or pools when running ADCC or other lysis-based assays. This kit contains exogenous EA as well as EFC substrate to allow for detection of lysis-based release of ProLabel-tagged KILR reporter protein. This kit contains all components required to assay 2 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-97-0001s.
Catalog No. NC1000669 Supplier Eurofins Discoverx Supplier No. 970001S
Add to Cart
Add to Cart
The KILR Detection Kit is a detection kit intended for use with KILR cell lines or pools when running ADCC or other lysis-based assays. This kit contains exogenous EA as well as EFC substrate to allow for detection of lysis-based release of ProLabel-tagged KILR reporter protein. This kit contains all components required to assay 2 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-97-0001s.