Learn More
Eurofins Discoverx KILR MM-1R Cell Pool, 2 vials. (97-1045P052)
Catalog No. 502004763
Encompass
The KILR MM-1R cell pool is a stable, engineered target cell pool, which closely reflects the heterogeneous native protein expression of tumor environments, used to measure direct cell death mediated by immunooncology treatments (e.g. checkpoint modulators, bispecifics, CAR-T, etc.) via Antibody Dependent Cell-mediated Cytotoxicity (ADCC), Complement Dependent Cytotoxicity (CDC), and Antibody Dependent Cell Phagocytosis (ADCP). This product uses EFC technology. The cell pool is a heterogeneous population of MM-1R cells that overexpress a ProLabel-tagged housekeeping gene. Upon effector cells-mediated cell lysis, ProLabel is released to the media, where EA in the detection kit produces an EFC signal proportional to the level of ProLabel released. This allows for calculation of percent lysis of the KILR cell pool. This stable cell pool is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kilr.
Catalog No. 50-200-4763 Supplier Eurofins Discoverx Supplier No. 971045P052
Add to Cart
Add to Cart
Request a Quote
The KILR MM-1R cell pool is a stable, engineered target cell pool, which closely reflects the heterogeneous native protein expression of tumor environments, used to measure direct cell death mediated by immunooncology treatments (e.g. checkpoint modulators, bispecifics, CAR-T, etc.) via Antibody Dependent Cell-mediated Cytotoxicity (ADCC), Complement Dependent Cytotoxicity (CDC), and Antibody Dependent Cell Phagocytosis (ADCP). This product uses EFC technology. The cell pool is a heterogeneous population of MM-1R cells that overexpress a ProLabel-tagged housekeeping gene. Upon effector cells-mediated cell lysis, ProLabel is released to the media, where EA in the detection kit produces an EFC signal proportional to the level of ProLabel released. This allows for calculation of percent lysis of the KILR cell pool. This stable cell pool is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kilr.