Learn More
Eurofins Discoverx KILR Retroparticles for Adherent Cells (Hygromycin B), 4 vials x 0.5. (97-0005)
Catalog No. 502004583
Encompass
The KILR Retroparticles for Adherent Cells (Hygromycin B) are MMLV particles designed to transduce adherent cells. This allows production of ProLabel-tagged, hygromycin B-resistant target cells of interest for use in cytotoxicity assays involving detection of cell lysis. This product uses EFC technology. The retroparticles mix can be used to transduce target cells with a ProLabel-tagged housekeeping gene. Target cell death in a cytotoxicity assay will release ProLabel to the media, where EA contained in the detection reagent will produce EFC signal. These retroparticles ship as 4 vials of cryopreserved retroparticles. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/virus/kilr/retroparticles-for-adherent-cells-(hygrom-97-0005.
Catalog No. 50-200-4583 Supplier Eurofins Discoverx Supplier No. 970005
The KILR Retroparticles for Adherent Cells (Hygromycin B) are MMLV particles designed to transduce adherent cells. This allows production of ProLabel-tagged, hygromycin B-resistant target cells of interest for use in cytotoxicity assays involving detection of cell lysis. This product uses EFC technology. The retroparticles mix can be used to transduce target cells with a ProLabel-tagged housekeeping gene. Target cell death in a cytotoxicity assay will release ProLabel to the media, where EA contained in the detection reagent will produce EFC signal. These retroparticles ship as 4 vials of cryopreserved retroparticles. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/virus/kilr/retroparticles-for-adherent-cells-(hygrom-97-0005.