Learn More
Enzo Life Sciences KIM-1 (human) monoclonal antibody (3F4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI9059060100
Encompass_Preferred
Clone: 3F4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human KIM-1. UniProt ID: Q96D42.2. Source: Purified from mouse ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7081