Learn More
BioVendor, LLC Kim-1 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL006R
Encompass
The urinary kidney injury molecule 1 (KIM-1) participates in renal tissue damage and repair and is proposed as a biomarker of early and subclinical AKI. KIM-1 is also elevated in the urine of a significant fraction of patients apparently recovered from an AKI. Besides its potential utility in the early and subclinical diagnosis of renal damage, this study suggests a new application of urinary KIM-1 in the non-invasive follow-up of renal repair after AKI. One Kit (96 wells). Type: sandwich ELISA. Sample Type: serum, urine. Sample requirement: 50 ¿l/well. Calibration range: 15.6-1000pg/ml. Limit of detection: 7.8pg/ml. Other names: Hepatitis A Virus Cellular Receptor 1, HAVcr-1, Kidney injury molecule 1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIMD-1, T-cell immunoglobulin mucin receptor 1, TIM, TIM-1, T-cell membrane protein 1, HAVCR1.
Catalog No. 50-240-6355