Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KIMAX™ Glass Bottle Reservoir
SDP

Catalog No. 12141412 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-141-412 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-141-412 Supplier DWK Life Sciences Supplier No. 17224F1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ KIMAX™ Glass Bottle Reservoir is manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements. Designed as a replacement reservoir for KIMAX™ buret assemblies.

  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Conforms to USP Type I and ASTM E438, Type I, Class A requirements
  • Designed as a replacement bottle reservoir for KIMAX buret assemblies
  • Supplied by DWK Life Sciences under the Kimble KIMAX brand
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 L
Color Clear
Includes White enamel marking spot
Material Glass
Sterility Non-sterile
Closure Included No
Graduated No
Mouth Narrow
Chemical Label Unlabeled
Autoclavable Autoclavable
Capacity (English) 34 oz.
Shape Round
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.