Learn More
DWK Life Sciences Kimble™ KIMAX™ Snap Caps for 300mL BOD Bottles
SDP
Catalog No. 02939 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Application):
Holds stopper securely in bottle and reduces evaporation of water seal during incubation
Catalog No. For Use With (Application)
02-939 Holds stopper securely in bottle and reduces evaporation of water seal during incubation
1 options
Catalog No. 02-939 Supplier DWK Life Sciences Supplier No. 15070L99
Holds stopper securely in bottle and reduces evaporation of water seal during incubation.
  • Case of 50
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Holds stopper securely in bottle and reduces evaporation of water seal during incubation
For Use With (Equipment) For 300mL BOD bottle