Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™-35 300mL BOD Bottles
SDP

Catalog No. p-207919 Shop All DWK Life Sciences Products
Change view
Click to view available options
No. Sequence:
1 to 24
Unnumbered
Stopper:
With stopper
Without stopper
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. Sequence Stopper
02-885B 1 to 24 With stopper
02-885A Unnumbered With stopper
02-940-1A Unnumbered Without stopper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-885B Supplier DWK Life Sciences Supplier No. 1507001

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis

Flared mouth forms a water seal to prevent air from being drawn into the bottle during incubation, while shoulder radius provides an interior shape that sweeps entrapped air out of the stopper opening.

  • Type 1, Class B borosilicate glass
  • Available numbered and unnumbered With glass stopper unless otherwise noted
  • Optional snap lid secures stopper in bottle and reduces evaporation of water seal during incubation (sold separately, 02-939)
  • Size: 2.75 dia. × 6.5 in. H (69 × 165mm); neck I.D. approximately 0.66 in.  (16.7mm)

Order Info

Plastic snap lids not included. For these, see Cat. No. 02-939.

Compliance

Conforms to ASTM™ E 438

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM E438 type I, class A
Volume (Metric) 300 mL
For Use With (Application) For sampling and incubation of aqueous samples
Material Borosilicate Glass
No. Sequence 1 to 24
Stopper With stopper
Volume (English) 10.14 oz.
Height (English) 6.5 in.
Height (Metric) 16.5 cm
Diameter (English) 2.7 in.
Diameter (Metric) 6.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.