Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Reagent Bottles
SDP

Catalog No. p-45238 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
20,000 mL
2000 mL
Diameter (English) Inner Neck:
1.06 in.
2.28 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Diameter (English) Inner Neck
02-886-8D 1000 mL 1.06 in.
02-886-8E 2000 mL 1.06 in.
02-886-8G 10,000 mL 2.28 in.
02-886-8H 20,000 mL 2.28 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-886-8D Supplier DWK Life Sciences Supplier No. 150931000
Only null left
Add to Cart
Add to Cart

For accurate rubber stopper fit

  • Borosilicate glass
  • Tooled top and narrow mouth

Order Info

Stoppers are available separately; order online

Compliance

Meet ASTM™ E 438-92, Type I, Class A requirements.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 33.81 oz., 1000mL
Capacity (Metric) 1000 mL
Capacity (English) 33.81 oz.
Diameter (English) 4.33 in.
Diameter (English) Inner Neck 1.06 in.
Diameter (Metric) Inner Neck 27 mm
Includes Tooled neck
Material Borosilicate Glass
Stopper Number 6
Certifications/Compliance ASTM E438-92, Type I, Class A
Mouth Narrow
Height (Metric) 200 mm
Height (English) 7.87 in.
Closure Material Rubber
Diameter (Metric) 110 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.