Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ KIMAX™ Squibb Pear-Shaped Separatory Funnels
SDPSCI_ED

Catalog No. S34695 Shop All DWK Life Sciences Products
Change view
Click to view available options
Stopcock:
2; PTFE
4; PTFE
Stopper Number:
22
Quantity:
2 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopcock Stopper Number Quantity
S34695 4; PTFE 22 2 Pack
S34694 2; PTFE 22 2 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S34695 Supplier Kimble™ Supplier No. 29048F250/EMD
This item is not returnable. View return policy

Permit the separation of lower layers of immiscible liquids.

  • Supplied with a ground glass stopper and a PTFE stopcock PTFE stopcocks are less likely to freeze than glass stopcocks

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Material Borosilicate Glass
Certifications/Compliance ASTM E1096, Type IV
For Use With (Application) Eliminates contamination
Length (Metric) Stem 6cm
Quantity 2 Pack
Stopcock 4; PTFE
Stopper Material Glass
Stopper Number 22
Product Type Squibb separatory funnel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.