Learn More
Sigma Aldrich Fine Chemicals Biosciences KimaxR centrifuge tube cap
Catalog No. 502905065
Encompass
Heavy-wall not graduated. Supplied with 24-410 screw-cap with a cemented-in rubber liner. Autoclavable.
Catalog No. 50-290-5065 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z25485112EA
Heavy-wall not graduated. Supplied with 24-410 screw-cap with a cemented-in rubber liner. Autoclavable.