Learn More
Sigma Aldrich Fine Chemicals Biosciences KimaxR centrifuge tube oil1EA
Catalog No. 502901606
Encompass
Used in the determination of residues in Liquefied Petroleum (LP) gases ASTM Method D2158. Certified to contain. Scale and legend are durable white ceramic enamel.
Catalog No. 50-290-1606 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2522631EA
Used in the determination of residues in Liquefied Petroleum (LP) gases ASTM Method D2158. Certified to contain. Scale and legend are durable white ceramic enamel.