Learn More
Sigma Aldrich Fine Chemicals Biosciences KimaxR heavy-duty wide-mou
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z5016626EA
Wall thickness and weight are at least two times greater than that of standard volumetric flasks for durability. Calibrated to contain. Meets ASTM E288 Class A unserialized requirements. Easy pipette access. Supplied with glass stopper.